Product Details

SNP ID
rs573839746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75815190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAATGAGCGCTCCGGGTCTCAGCG[C/G]GGTGGAGGGGTTGCACTGCGGTAAT
Phenotype
MIM: 602493
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100506113 PubMed Links

Gene Details

Gene
LOC100506113
Gene Name
uncharacterized LOC100506113
There are no transcripts associated with this gene.

Gene
UVRAG
Gene Name
UV radiation resistance associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003369.3 339 UTR 5 NP_003360.2
XM_011545226.2 339 UTR 5 XP_011543528.1
XM_011545227.2 339 UTR 5 XP_011543529.1
XM_011545228.2 339 UTR 5 XP_011543530.1
XM_017018224.1 339 Intron XP_016873713.1
XM_017018225.1 339 Intron XP_016873714.1
XM_017018226.1 339 UTR 5 XP_016873715.1
XM_017018227.1 339 Intron XP_016873716.1

View Full Product Details