Product Details

SNP ID
rs79589438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:50747206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTCCTAAAGACCCTTTCAGAAT[A/G]TATGCATTCCTCTTTTCACGATTGT
Phenotype
MIM: 605018
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYLD PubMed Links

Gene Details

Gene
CYLD
Gene Name
CYLD lysine 63 deubiquitinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042355.1 Intron NP_001035814.1
NM_001042412.1 Intron NP_001035877.1
NM_015247.2 Intron NP_056062.1
XM_005255812.2 Intron XP_005255869.1
XM_006721149.1 Intron XP_006721212.1
XM_011522907.2 Intron XP_011521209.1
XM_011522908.1 Intron XP_011521210.1
XM_011522911.2 Intron XP_011521213.1
XM_017022977.1 Intron XP_016878466.1
XM_017022978.1 Intron XP_016878467.1
XM_017022979.1 Intron XP_016878468.1
XM_017022980.1 Intron XP_016878469.1
XM_017022981.1 Intron XP_016878470.1
Gene
MIR3181
Gene Name
microRNA 3181
There are no transcripts associated with this gene.

View Full Product Details