Product Details
- SNP ID
-
rs115838884
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:58635228 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTGGCAACAGAAGGGAGCAGTAGG[C/T]GATCACTTTCAACTGCTTCCGGAAT
- Phenotype
-
MIM: 607861
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DACT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148419855] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DACT1
- Gene Name
- dishevelled binding antagonist of beta catenin 1
View Full Product Details