Product Details

SNP ID
rs9341801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:80005446 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCAAGTAAAGCAGGCGAATAAAG[A/G]AATTGTTATTCCCATTGCCAAACGA
Phenotype
MIM: 604092
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTK PubMed Links

Gene Details

Gene
TTK
Gene Name
TTK protein kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166691.1 Intron NP_001160163.1
NM_003318.4 Intron NP_003309.2
XM_011536099.2 Intron XP_011534401.1
XM_011536100.2 Intron XP_011534402.1
XM_017011242.1 Intron XP_016866731.1

View Full Product Details