Product Details

SNP ID
rs3788922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:108442312 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGGTAAATGATTTCTTTATCTCT[C/G]AAATCATGCCAAATTGGTACATCAC
Phenotype
MIM: 303630 MIM: 303631
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COL4A5 PubMed Links

Gene Details

Gene
COL4A5
Gene Name
collagen type IV alpha 5 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000495.4 Intron NP_000486.1
NM_033380.2 Intron NP_203699.1
XM_011530849.2 Intron XP_011529151.2
XM_017029259.1 Intron XP_016884748.1
XM_017029260.1 Intron XP_016884749.1
XM_017029261.1 Intron XP_016884750.1
XM_017029262.1 Intron XP_016884751.1
XM_017029263.1 Intron XP_016884752.1
Gene
COL4A6
Gene Name
collagen type IV alpha 6 chain
There are no transcripts associated with this gene.

View Full Product Details