Product Details

SNP ID
rs976953689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:96502496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTTGGACCCAGGGCTGAGGCAG[A/G]CCCCCCCCTCCCTCCCGCCTCAGTG
Phenotype
MIM: 605553
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC730202 PubMed Links

Gene Details

Gene
LOC730202
Gene Name
uncharacterized LOC730202
There are no transcripts associated with this gene.

Gene
PAPOLA
Gene Name
poly(A) polymerase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252006.1 121 UTR 5 NP_001238935.1
NM_001252007.1 121 UTR 5 NP_001238936.1
NM_001293627.1 121 UTR 5 NP_001280556.1
NM_001293628.1 121 Intron NP_001280557.1
NM_001293632.1 121 UTR 5 NP_001280561.1
NM_032632.4 121 UTR 5 NP_116021.2

View Full Product Details