Product Details

SNP ID
hCV27840414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:9850183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTCTTTCTTTTCAAGTCAAATA[C/T]AACGACTAATGTCCCAGACCCTGTT
Phenotype
MIM: 607758
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTNNBIP1 PubMed Links

Gene Details

Gene
CTNNBIP1
Gene Name
catenin beta interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012329.1 1088 UTR 3 NP_001012329.1
NM_020248.2 1088 UTR 3 NP_064633.1

View Full Product Details