Product Details

SNP ID
rs377644323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58453273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGCCTGGAAATGTCAGGCAAGC[A/G]CAGCAGGGGAAGGTCTCAGTGATGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF324B PubMed Links

Gene Details

Gene
ZNF324B
Gene Name
zinc finger protein 324B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207395.2 Intron NP_997278.2
XM_005258915.4 Intron XP_005258972.1
XM_005258918.4 Intron XP_005258975.1

View Full Product Details