Product Details

SNP ID
rs146431654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56643689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACCGGCCCGCCTGTTTTTTCGCC[A/T]GTGGCTTAGGAACTCCAGCCTCACC
Phenotype
MIM: 156350 MIM: 156349 MIM: 156356 MIM: 156357
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MT1A PubMed Links

Gene Details

Gene
MT1A
Gene Name
metallothionein 1A
There are no transcripts associated with this gene.

Gene
MT1B
Gene Name
metallothionein 1B
There are no transcripts associated with this gene.

Gene
MT1DP
Gene Name
metallothionein 1D, pseudogene
There are no transcripts associated with this gene.

Gene
MT1JP
Gene Name
metallothionein 1J, pseudogene
There are no transcripts associated with this gene.

Gene
MT1M
Gene Name
metallothionein 1M
There are no transcripts associated with this gene.

View Full Product Details