Product Details

SNP ID
rs980120935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58420324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGACTTTATTATACCTCTTGCCA[C/T]TTTGCCCAAGTCATTAATGACAGCT
Phenotype
MIM: 612482
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSF5 PubMed Links

Gene Details

Gene
HSF5
Gene Name
heat shock transcription factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080439.2 3954 UTR 3 NP_001073908.2
Gene
RNF43
Gene Name
ring finger protein 43
There are no transcripts associated with this gene.

View Full Product Details