Product Details

SNP ID
rs146607841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:116855493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGCATCAGGAAGAATACTCGC[A/G]GAGAGGTTTGGTAGAAACTAGAGTC
Phenotype
MIM: 610701
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENO4 PubMed Links

Gene Details

Gene
ENO4
Gene Name
enolase family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242699.1 Intron NP_001229628.1
XM_005269815.3 Intron XP_005269872.1
XM_006717835.3 Intron XP_006717898.1
XM_006717836.3 Intron XP_006717899.1
XM_011539797.2 Intron XP_011538099.1
XM_011539799.2 Intron XP_011538101.1
XM_017016241.1 Intron XP_016871730.1
Gene
HSPA12A
Gene Name
heat shock protein family A (Hsp70) member 12A
There are no transcripts associated with this gene.

View Full Product Details