Product Details

SNP ID
rs59493015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61392608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGTTTCCGGCAGCGCCGCGCTG[C/G]TCCGGGAGCCGCTGTGGCAGCGTAT
Phenotype
MIM: 613277
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM216 PubMed Links

Gene Details

Gene
TMEM216
Gene Name
transmembrane protein 216
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173990.2 922 UTR 5 NP_001167461.1
NM_001173991.2 922 UTR 5 NP_001167462.1
NM_016499.5 922 UTR 5 NP_057583.2
XM_005274039.3 922 UTR 5 XP_005274096.1
XM_017017882.1 922 UTR 5 XP_016873371.1

View Full Product Details