Product Details

SNP ID
rs72838317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4988392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGCACCACTAGCCTCTCGGCAT[C/T]GGTGGTTTCTCCTTACTCTTCAGAC
Phenotype
MIM: 611508 MIM: 603060
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAMTA2 PubMed Links

Gene Details

Gene
CAMTA2
Gene Name
calmodulin binding transcription activator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171166.1 1155 Intron NP_001164637.1
NM_001171167.1 1155 Intron NP_001164638.1
NM_001171168.1 1155 Intron NP_001164639.1
NM_015099.3 1155 Intron NP_055914.2
XM_006721478.3 1155 Intron XP_006721541.1
XM_006721481.3 1155 Intron XP_006721544.1
XM_006721482.3 1155 Intron XP_006721545.1
XM_011523746.2 1155 Intron XP_011522048.1
XM_011523747.2 1155 Intron XP_011522049.1
XM_011523748.2 1155 Intron XP_011522050.1
XM_011523749.2 1155 Intron XP_011522051.1
Gene
INCA1
Gene Name
inhibitor of CDK, cyclin A1 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167985.1 1155 UTR 3 NP_001161457.1
NM_001167986.1 1155 UTR 3 NP_001161458.1
NM_001167987.1 1155 UTR 3 NP_001161459.1
NM_213726.2 1155 UTR 3 NP_998891.2
XM_005256627.3 1155 UTR 3 XP_005256684.1
XM_005256628.4 1155 UTR 3 XP_005256685.1
XM_005256629.1 1155 UTR 3 XP_005256686.1
XM_006721517.1 1155 UTR 3 XP_006721580.1
XM_006721518.2 1155 UTR 3 XP_006721581.1
XM_006721519.3 1155 UTR 3 XP_006721582.1
XM_011523832.2 1155 UTR 3 XP_011522134.1
XM_011523834.2 1155 Intron XP_011522136.1
Gene
KIF1C
Gene Name
kinesin family member 1C
There are no transcripts associated with this gene.

Gene
LOC101927979
Gene Name
translation initiation factor IF-2
There are no transcripts associated with this gene.

View Full Product Details