Product Details

SNP ID
rs16980628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15615534 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCTCCCTCCATGCTCTTAACCAT[G/T]TTTACCCATCATTGGTCCTGGAGCT
Phenotype
MIM: 611545
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP4F8 PubMed Links

Gene Details

Gene
CYP4F8
Gene Name
cytochrome P450 family 4 subfamily F member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007253.3 Intron NP_009184.1
XM_017026232.1 Intron XP_016881721.1

View Full Product Details