Product Details

SNP ID
rs74491678
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:15760691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCCAAGGCTGCAGCCTCTAGCT[A/G]GTCTGGGTGGCTGGCTGGATCCCTG
Phenotype
MIM: 607747
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBLIM1 PubMed Links

Gene Details

Gene
FBLIM1
Gene Name
filamin binding LIM protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024215.1 Intron NP_001019386.1
NM_001024216.1 Intron NP_001019387.1
NM_017556.2 Intron NP_060026.2
XM_005245900.1 Intron XP_005245957.1
XM_005245901.1 Intron XP_005245958.1
XM_005245902.1 Intron XP_005245959.1
XM_005245903.1 Intron XP_005245960.1
XM_005245909.3 Intron XP_005245966.1
XM_006710704.3 Intron XP_006710767.1
XM_006710705.1 Intron XP_006710768.1
XM_011541616.2 Intron XP_011539918.1
XM_011541617.2 Intron XP_011539919.1
XM_011541618.2 Intron XP_011539920.1
XM_017001519.1 Intron XP_016857008.1
XM_017001520.1 Intron XP_016857009.1
XM_017001521.1 Intron XP_016857010.1
XM_017001522.1 Intron XP_016857011.1
XM_017001523.1 Intron XP_016857012.1
XM_017001524.1 Intron XP_016857013.1
XM_017001525.1 Intron XP_016857014.1
XM_017001526.1 Intron XP_016857015.1

View Full Product Details