Product Details

SNP ID
rs533953547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:69573830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAGCGCCAGAACGGGGATGGGG[G/T]TGGGGCGGACACTCGTGTTCAGAGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC177 PubMed Links

Gene Details

Gene
CCDC177
Gene Name
coiled-coil domain containing 177
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271507.1 Intron NP_001258436.1

View Full Product Details