Product Details

SNP ID
hCV27843145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113948357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAGCCTGGTGTGTCAGCCGCTG[A/C]TAGCGGGCATATGCTTCGCCTTTCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GRAMD1C PubMed Links

Gene Details

Gene
GRAMD1C
Gene Name
GRAM domain containing 1C
There are no transcripts associated with this gene.

Gene
ZDHHC23
Gene Name
zinc finger DHHC-type containing 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320466.1 Intron NP_001307395.1
NM_001320467.1 Intron NP_001307396.1
NM_001320468.1 Intron NP_001307397.1
NM_173570.4 Intron NP_775841.2

View Full Product Details