Product Details

SNP ID
rs190635505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123498053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACACAATATTAAATAAACATCT[C/T]TGCATGTAATTGGTCTAACTTTATG
Phenotype
MIM: 606604
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO32 PubMed Links

Gene Details

Gene
FBXO32
Gene Name
F-box protein 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242463.1 6262 UTR 3 NP_001229392.1
NM_058229.3 6262 UTR 3 NP_478136.1
NM_148177.2 6262 UTR 3 NP_680482.1

View Full Product Details