Product Details

SNP ID
rs971090943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20343364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGGACATGGGAGTGGAGTGACA[A/G]GACGCACTCAGCTCGTGGCCCCACT
Phenotype
MIM: 607725 MIM: 608513
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PARP2 PubMed Links

Gene Details

Gene
PARP2
Gene Name
poly(ADP-ribose) polymerase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042618.1 Intron NP_001036083.1
NM_005484.3 Intron NP_005475.2
Gene
RPPH1
Gene Name
ribonuclease P RNA component H1
There are no transcripts associated with this gene.

View Full Product Details