Product Details

SNP ID
rs73297453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73715754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGTATTCCTCCTTCATCTTAACA[G/T]CTGGCACATGAGAAGAAAGAGGTTA
Phenotype
MIM: 604010
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ELMSAN1 PubMed Links

Gene Details

Gene
ELMSAN1
Gene Name
ELM2 and Myb/SANT domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001043318.2 7089 UTR 3 NP_001036783.1
NM_194278.3 7089 UTR 3 NP_919254.2
XM_005268204.1 7089 UTR 3 XP_005268261.1
XM_005268205.1 7089 UTR 3 XP_005268262.1
XM_005268206.1 7089 UTR 3 XP_005268263.1
Gene
PNMA1
Gene Name
paraneoplastic Ma antigen 1
There are no transcripts associated with this gene.

View Full Product Details