Product Details

SNP ID
rs112323844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122577300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAAATATGGGTGTGTAGGGAGAA[C/T]TGGGGGGAAAGGAGTGGGATGCCCA
Phenotype
MIM: 613143 MIM: 612066
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DTX3L PubMed Links

Gene Details

Gene
DTX3L
Gene Name
deltex E3 ubiquitin ligase 3L
There are no transcripts associated with this gene.

Gene
PARP15
Gene Name
poly(ADP-ribose) polymerase family member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113523.2 1371 Intron NP_001106995.1
NM_001308320.1 1371 Intron NP_001295249.1
NM_001308321.1 1371 Intron NP_001295250.1
NM_152615.2 1371 Intron NP_689828.1
XM_005247159.3 1371 UTR 5 XP_005247216.1
XM_005247160.3 1371 UTR 5 XP_005247217.1
XM_011512475.2 1371 UTR 5 XP_011510777.1
XM_011512476.2 1371 UTR 5 XP_011510778.1
XM_011512477.2 1371 UTR 5 XP_011510779.1
XM_011512478.2 1371 Intron XP_011510780.1
XM_011512479.2 1371 Intron XP_011510781.1
XM_011512480.2 1371 UTR 5 XP_011510782.1
XM_017005791.1 1371 UTR 5 XP_016861280.1
XM_017005792.1 1371 Intron XP_016861281.1

View Full Product Details