Product Details

SNP ID
rs115332130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:127730469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTACGATGCTGCAGCGGTTTTCC[A/G]GTTTTCCGCTTCCCTTCATCGTAGC
Phenotype
MIM: 610796
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A31 PubMed Links

Gene Details

Gene
SLC25A31
Gene Name
solute carrier family 25 member 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318467.1 92 UTR 5 NP_001305396.1
NM_031291.3 92 UTR 5 NP_112581.1
XM_011532298.1 92 UTR 5 XP_011530600.1

View Full Product Details