Product Details

SNP ID
rs76798065
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:37716171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTAGAGGGTTCCCAGAGAGCCAT[C/T]GCCCCACATCACATCTGTGCAGGGC
Phenotype
MIM: 609977 MIM: 611123
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDCA8 PubMed Links

Gene Details

Gene
CDCA8
Gene Name
cell division cycle associated 8
There are no transcripts associated with this gene.

Gene
EPHA10
Gene Name
EPH receptor A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099439.1 5314 UTR 3 NP_001092909.1
NM_173641.2 5314 Intron NP_775912.2
XM_017001080.1 5314 Intron XP_016856569.1
XM_017001081.1 5314 Intron XP_016856570.1

View Full Product Details