Product Details

SNP ID
rs115766017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:121801378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCGCAAGGAAAGATCGGGTTCC[A/G]TTTTTCCCGCGGATTCTGGTGCCTG
Phenotype
MIM: 606180
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EXOSC9 PubMed Links

Gene Details

Gene
EXOSC9
Gene Name
exosome component 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034194.1 62 UTR 5 NP_001029366.1
NM_005033.2 62 UTR 5 NP_005024.2
XM_011532035.2 62 UTR 5 XP_011530337.1

View Full Product Details