Product Details

SNP ID
rs58598489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:133080967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGCAACGAAAGGAGCCCTCCCGG[C/T]CTGCGCCGGATGGCCCCGGGCGGTG
Phenotype
MIM: 604082
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF140 PubMed Links

Gene Details

Gene
ZNF140
Gene Name
zinc finger protein 140
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300776.1 765 UTR 5 NP_001287705.1
NM_001300777.1 765 UTR 5 NP_001287706.1
NM_001300778.1 765 UTR 5 NP_001287707.1
NM_003440.3 765 UTR 5 NP_003431.2
XM_005266188.4 765 UTR 5 XP_005266245.1
XM_011534833.2 765 UTR 5 XP_011533135.1
XM_011534834.2 765 Intron XP_011533136.1
XM_011534835.2 765 Intron XP_011533137.1
XM_011534836.2 765 Intron XP_011533138.1
XM_011534837.2 765 Intron XP_011533139.1
XM_011534838.2 765 UTR 5 XP_011533140.1
XM_011534839.2 765 UTR 5 XP_011533141.1
XM_011534840.2 765 Intron XP_011533142.1
XM_017019924.1 765 UTR 5 XP_016875413.1
XM_017019925.1 765 Intron XP_016875414.1

View Full Product Details