Product Details

SNP ID
hCV27846159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20781498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACTTGTGAAAGAAAATGGGATA[C/T]GCCAATCAAAGGAGAGGTCATATTT
Phenotype
MIM: 601981
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNASE6 PubMed Links

Gene Details

Gene
RNASE6
Gene Name
ribonuclease A family member k6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005615.4 Intron NP_005606.1

View Full Product Details