Product Details

SNP ID
rs376510543
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:54226415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGAAATCAAGCTAGAACAAAC[A/G]CGTCCCCACTCCTCCCCTGCCTGTT
Phenotype
MIM: 611843 MIM: 607390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL37 PubMed Links

Gene Details

Gene
MRPL37
Gene Name
mitochondrial ribosomal protein L37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016491.3 2210 Intron NP_057575.2
XM_005270920.3 2210 Intron XP_005270977.1
Gene
SSBP3
Gene Name
single stranded DNA binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009955.3 2210 UTR 3 NP_001009955.1
NM_018070.4 2210 UTR 3 NP_060540.2
NM_145716.3 2210 UTR 3 NP_663768.1
XM_006710545.3 2210 Intron XP_006710608.1
XM_017000897.1 2210 Intron XP_016856386.1
XM_017000898.1 2210 Intron XP_016856387.1
XM_017000899.1 2210 Intron XP_016856388.1

View Full Product Details