Product Details

SNP ID
rs187032702
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:36877294 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTAGCGGCCCCAGGTCTCTTC[C/T]ACAGGCGAGTCTAGAGTTCGCTCCT
Phenotype
MIM: 608667
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NIPBL PubMed Links

Gene Details

Gene
NIPBL
Gene Name
NIPBL, cohesin loading factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015384.4 Intron NP_056199.2
NM_133433.3 Intron NP_597677.2
XM_005248280.3 Intron XP_005248337.1
XM_005248282.4 Intron XP_005248339.3
XM_006714467.2 Intron XP_006714530.1
XM_006714468.2 Intron XP_006714531.1
XM_011514015.1 Intron XP_011512317.1
XM_017009329.1 Intron XP_016864818.1
XM_017009330.1 Intron XP_016864819.1
XM_017009331.1 Intron XP_016864820.1
Gene
NIPBL-AS1
Gene Name
NIPBL antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details