Product Details

SNP ID
rs184331529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124117360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTGAAATCTTTGTCTCCTTTTT[G/T]TTAAAGGTTCCAACTTCCTAACTCC
Phenotype
MIM: 602929
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VWA5A PubMed Links

Gene Details

Gene
VWA5A
Gene Name
von Willebrand factor A domain containing 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130142.1 Intron NP_001123614.1
NM_014622.4 Intron NP_055437.2
NM_198315.2 Intron NP_938057.1
XM_011542828.2 Intron XP_011541130.1

View Full Product Details