Product Details

SNP ID
rs4346380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241323808 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCAAACAGAATTATAACATTTTC[A/G]GAACTAAGGCTCATTTAAAGATACA
Phenotype
MIM: 142695 MIM: 601506
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HDLBP PubMed Links

Gene Details

Gene
HDLBP
Gene Name
high density lipoprotein binding protein
There are no transcripts associated with this gene.

Gene
SEPT2
Gene Name
septin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008491.2 Intron NP_001008491.1
NM_001008492.2 Intron NP_001008492.1
NM_001282972.1 Intron NP_001269901.1
NM_001282973.1 Intron NP_001269902.1
NM_001321029.1 Intron NP_001307958.1
NM_001321030.1 Intron NP_001307959.1
NM_001321031.1 Intron NP_001307960.1
NM_001321032.1 Intron NP_001307961.1
NM_001321033.1 Intron NP_001307962.1
NM_001321034.1 Intron NP_001307963.1
NM_001321035.1 Intron NP_001307964.1
NM_004404.4 Intron NP_004395.1
NM_006155.2 Intron NP_006146.1
XM_011511237.1 Intron XP_011509539.1
XM_011511238.1 Intron XP_011509540.1
XM_011511240.1 Intron XP_011509542.1
XM_017004189.1 Intron XP_016859678.1
XM_017004190.1 Intron XP_016859679.1
XM_017004191.1 Intron XP_016859680.1
XM_017004192.1 Intron XP_016859681.1
XM_017004193.1 Intron XP_016859682.1
XM_017004194.1 Intron XP_016859683.1
XM_017004195.1 Intron XP_016859684.1
XM_017004196.1 Intron XP_016859685.1
XM_017004197.1 Intron XP_016859686.1
XM_017004198.1 Intron XP_016859687.1
XM_017004199.1 Intron XP_016859688.1
XM_017004200.1 Intron XP_016859689.1
XM_017004201.1 Intron XP_016859690.1
XM_017004202.1 Intron XP_016859691.1
XM_017004203.1 Intron XP_016859692.1
XM_017004204.1 Intron XP_016859693.1
XM_017004205.1 Intron XP_016859694.1
XM_017004206.1 Intron XP_016859695.1
XM_017004207.1 Intron XP_016859696.1
XM_017004208.1 Intron XP_016859697.1
XM_017004209.1 Intron XP_016859698.1
XM_017004210.1 Intron XP_016859699.1
XM_017004211.1 Intron XP_016859700.1
XM_017004212.1 Intron XP_016859701.1
XM_017004213.1 Intron XP_016859702.1

View Full Product Details