Product Details

SNP ID
rs142499430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64550943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGTTGGGATTACGATTAGGGA[A/T]GTGGTACTGGAAAGATGAAACCAGA
Phenotype
MIM: 140560
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HSPA2 PubMed Links

Gene Details

Gene
HSPA2
Gene Name
heat shock protein family A (Hsp70) member 2
There are no transcripts associated with this gene.

Gene
PPP1R36
Gene Name
protein phosphatase 1 regulatory subunit 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172365.2 156 Missense Mutation AAG,ATG K31M NP_758953.1
XM_005267354.3 156 Missense Mutation AAG,ATG K19M XP_005267411.1
XM_005267355.3 156 Intron XP_005267412.1
XM_011536468.2 156 Missense Mutation AAG,ATG K31M XP_011534770.1

View Full Product Details