Product Details
- SNP ID
-
hCV27847447
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:40525605 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCAGGTGGAAGATTGTCCTGAAGA[C/T]TAGGCAGAACAATTCATAAGAAGTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF620
PubMed Links
Gene Details
- Gene
- ZNF620
- Gene Name
- zinc finger protein 620
There are no transcripts associated with this gene.
- Gene
- ZNF621
- Gene Name
- zinc finger protein 621
View Full Product Details