Product Details
- SNP ID
-
rs907386409
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:43303104 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTCTTCTTATTTTGTAGATATCC[A/G]TGACGACATTGAAAATGAATTTTTT
- Phenotype
-
MIM: 612760
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SNRK
PubMed Links
Gene Details
- Gene
- SNRK
- Gene Name
- SNF related kinase
View Full Product Details