Product Details

SNP ID
rs59371147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68360529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGTGACTTACAGACCCTGTGAA[A/G]TGAGGGTTGGTTTTGTGATTAACCT
Phenotype
MIM: 610087 MIM: 605777
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRMT7 PubMed Links

Gene Details

Gene
PRMT7
Gene Name
protein arginine methyltransferase 7
There are no transcripts associated with this gene.

Gene
SMPD3
Gene Name
sphingomyelin phosphodiesterase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018667.3 3467 UTR 3 NP_061137.1
XM_005256031.3 3467 UTR 3 XP_005256088.1
XM_005256032.3 3467 UTR 3 XP_005256089.1
XM_011523207.1 3467 UTR 3 XP_011521509.1
XM_011523208.2 3467 UTR 3 XP_011521510.1
XM_011523209.2 3467 UTR 3 XP_011521511.1
XM_011523210.2 3467 Intron XP_011521512.1
XM_017023405.1 3467 UTR 3 XP_016878894.1
XM_017023406.1 3467 UTR 3 XP_016878895.1
XM_017023407.1 3467 UTR 3 XP_016878896.1
XM_017023408.1 3467 UTR 3 XP_016878897.1

View Full Product Details