Product Details

SNP ID
rs115255322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:83458566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTCATTGGCTCTAGTACCATCT[C/G]TGGATGTTACAAAACTCTCAGTTAC
Phenotype
MIM: 603583
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DLG2 PubMed Links

Gene Details

Gene
DLG2
Gene Name
discs large MAGUK scaffold protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142699.1 3698 UTR 3 NP_001136171.1
NM_001142700.1 3698 UTR 3 NP_001136172.1
NM_001142702.1 3698 UTR 3 NP_001136174.1
NM_001206769.1 3698 UTR 3 NP_001193698.1
NM_001300983.1 3698 UTR 3 NP_001287912.1
NM_001364.3 3698 UTR 3 NP_001355.2
XM_005273810.4 3698 UTR 3 XP_005273867.1
XM_005273811.4 3698 UTR 3 XP_005273868.1
XM_011544778.2 3698 UTR 3 XP_011543080.1
XM_011544780.2 3698 UTR 3 XP_011543082.1
XM_011544782.2 3698 UTR 3 XP_011543084.1
XM_011544788.2 3698 UTR 3 XP_011543090.1
XM_017017254.1 3698 UTR 3 XP_016872743.1
XM_017017255.1 3698 UTR 3 XP_016872744.1
XM_017017256.1 3698 UTR 3 XP_016872745.1
XM_017017257.1 3698 UTR 3 XP_016872746.1
XM_017017258.1 3698 UTR 3 XP_016872747.1
XM_017017259.1 3698 UTR 3 XP_016872748.1
XM_017017260.1 3698 UTR 3 XP_016872749.1
XM_017017261.1 3698 UTR 3 XP_016872750.1
XM_017017262.1 3698 UTR 3 XP_016872751.1
XM_017017263.1 3698 UTR 3 XP_016872752.1
XM_017017264.1 3698 UTR 3 XP_016872753.1
XM_017017265.1 3698 UTR 3 XP_016872754.1
XM_017017266.1 3698 UTR 3 XP_016872755.1
XM_017017267.1 3698 UTR 3 XP_016872756.1
XM_017017268.1 3698 UTR 3 XP_016872757.1
XM_017017269.1 3698 UTR 3 XP_016872758.1
XM_017017270.1 3698 UTR 3 XP_016872759.1
XM_017017271.1 3698 UTR 3 XP_016872760.1
XM_017017272.1 3698 UTR 3 XP_016872761.1
XM_017017273.1 3698 UTR 3 XP_016872762.1
XM_017017274.1 3698 UTR 3 XP_016872763.1
XM_017017275.1 3698 UTR 3 XP_016872764.1
XM_017017276.1 3698 UTR 3 XP_016872765.1
XM_017017277.1 3698 UTR 3 XP_016872766.1
XM_017017278.1 3698 UTR 3 XP_016872767.1
XM_017017279.1 3698 UTR 3 XP_016872768.1
XM_017017280.1 3698 UTR 3 XP_016872769.1
XM_017017281.1 3698 UTR 3 XP_016872770.1
XM_017017282.1 3698 Intron XP_016872771.1
XM_017017283.1 3698 UTR 3 XP_016872772.1
XM_017017284.1 3698 UTR 3 XP_016872773.1
XM_017017285.1 3698 UTR 3 XP_016872774.1
XM_017017286.1 3698 UTR 3 XP_016872775.1
XM_017017287.1 3698 UTR 3 XP_016872776.1
XM_017017288.1 3698 UTR 3 XP_016872777.1
XM_017017289.1 3698 UTR 3 XP_016872778.1
XM_017017290.1 3698 UTR 3 XP_016872779.1
XM_017017291.1 3698 UTR 3 XP_016872780.1
XM_017017292.1 3698 UTR 3 XP_016872781.1

View Full Product Details