Product Details

SNP ID
rs115171840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:78236227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGACCGAATCACCGCTCCTGAGCC[C/T]GGTGCGGGGCTGCCGCTATCGCCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP78 PubMed Links

Gene Details

Gene
CEP78
Gene Name
centrosomal protein 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098802.1 166 UTR 5 NP_001092272.1
NM_032171.1 166 UTR 5 NP_115547.1
XM_005252263.4 166 UTR 5 XP_005252320.1
XM_005252265.2 166 UTR 5 XP_005252322.1
XM_005252266.4 166 UTR 5 XP_005252323.1
XM_017015192.1 166 UTR 5 XP_016870681.1
XM_017015193.1 166 UTR 5 XP_016870682.1
XM_017015194.1 166 UTR 5 XP_016870683.1
XM_017015195.1 166 UTR 5 XP_016870684.1
XM_017015196.1 166 UTR 5 XP_016870685.1

View Full Product Details