Product Details

SNP ID
rs10305750
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:150811349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGAAAATACCTGGAAGATTTAA[C/T]TCCTTAGGACTCTTTGAAAAGTACA
Phenotype
MIM: 126110 MIM: 601105
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARNT PubMed Links

Gene Details

Gene
ARNT
Gene Name
aryl hydrocarbon receptor nuclear translocator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197325.1 3073 UTR 3 NP_001184254.1
NM_001286035.1 3073 UTR 3 NP_001272964.1
NM_001286036.1 3073 UTR 3 NP_001272965.1
NM_001668.3 3073 UTR 3 NP_001659.1
NM_178427.2 3073 UTR 3 NP_848514.1
XM_005245151.1 3073 UTR 3 XP_005245208.1
XM_005245153.1 3073 UTR 3 XP_005245210.1
XM_005245154.3 3073 UTR 3 XP_005245211.1
XM_005245157.1 3073 UTR 3 XP_005245214.1
XM_011509542.2 3073 UTR 3 XP_011507844.1
XM_011509543.2 3073 UTR 3 XP_011507845.1
XM_011509544.2 3073 UTR 3 XP_011507846.1
XM_011509545.2 3073 UTR 3 XP_011507847.1
XM_011509546.1 3073 UTR 3 XP_011507848.1
XM_011509547.2 3073 UTR 3 XP_011507849.1
XM_017001288.1 3073 UTR 3 XP_016856777.1
XM_017001289.1 3073 UTR 3 XP_016856778.1
XM_017001290.1 3073 UTR 3 XP_016856779.1
XM_017001291.1 3073 UTR 3 XP_016856780.1
XM_017001292.1 3073 UTR 3 XP_016856781.1
XM_017001293.1 3073 UTR 3 XP_016856782.1
XM_017001294.1 3073 UTR 3 XP_016856783.1
XM_017001295.1 3073 UTR 3 XP_016856784.1
XM_017001296.1 3073 UTR 3 XP_016856785.1
Gene
CTSK
Gene Name
cathepsin K
There are no transcripts associated with this gene.

View Full Product Details