Product Details

SNP ID
rs936339345
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:150622209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTGGGCTCTGTGGGGTTCCCCA[C/T]GCTCACGGCTCCTTTCTCCCACACT
Phenotype
MIM: 603061
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENSA PubMed Links

Gene Details

Gene
ENSA
Gene Name
endosulfine alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004436.2 1017 UTR 3 NP_004427.1
NM_207042.1 1017 UTR 3 NP_996925.1
NM_207043.1 1017 Intron NP_996926.1
NM_207044.1 1017 Intron NP_996927.1
NM_207045.1 1017 UTR 3 NP_996928.1
NM_207046.1 1017 UTR 3 NP_996929.1
NM_207047.1 1017 Intron NP_996930.1
NM_207168.1 1017 Intron NP_997051.1

View Full Product Details