Product Details

SNP ID
rs7080579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73744271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATGGGAAGATGAAAGAGGCGCA[A/T]GCGCGGCATCCGGGTCCCCGAAATG
Phenotype
MIM: 607185
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SEC24C PubMed Links

Gene Details

Gene
SEC24C
Gene Name
SEC24 homolog C, COPII coat complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004922.3 108 Intron NP_004913.2
NM_198597.2 108 Intron NP_940999.1
XM_011540380.2 108 Intron XP_011538682.1
XM_011540381.2 108 UTR 5 XP_011538683.1
XM_017016967.1 108 Intron XP_016872456.1
XM_017016968.1 108 UTR 5 XP_016872457.1

View Full Product Details