Product Details

SNP ID
rs145657384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30218300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCACTGCCAGTACCTCCTGTCAC[C/G]ATAGTAAGCCCAAGGAAGGGCTGAC
Phenotype
MIM: 300098
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAGEB2 PubMed Links

Gene Details

Gene
MAGEB2
Gene Name
MAGE family member B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002364.4 Intron NP_002355.2
XM_011545512.1 Intron XP_011543814.1

View Full Product Details