Product Details

SNP ID
rs189666845
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47561129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGACGTGACCCTGACCCAATAAG[A/G]GTGGAAGGCTGAGTCCCGCAGAGCC
Phenotype
MIM: 311010
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ARAF PubMed Links

Gene Details

Gene
ARAF
Gene Name
A-Raf proto-oncogene, serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256196.1 30 UTR 5 NP_001243125.1
NM_001256197.1 30 UTR 5 NP_001243126.1
NM_001654.4 30 UTR 5 NP_001645.1
XM_006724529.2 30 Intron XP_006724592.1
XM_011543906.2 30 Intron XP_011542208.1
XM_011543908.2 30 Intron XP_011542210.1
XM_011543909.2 30 Intron XP_011542211.1

View Full Product Details