Product Details

SNP ID
rs116869463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:86555560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTGCTCCTCATGTAGGACGCGGT[C/T]CCCTCCTCTGAGATCCTGCAGACTC
Phenotype
MIM: 616820
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLJ30679 PubMed Links

Gene Details

Gene
FLJ30679
Gene Name
uncharacterized protein FLJ30679
There are no transcripts associated with this gene.

Gene
FOXC2-AS1
Gene Name
FOXC2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
MTHFSD
Gene Name
methenyltetrahydrofolate synthetase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159377.1 Intron NP_001152849.1
NM_001159378.1 Intron NP_001152850.1
NM_001159379.1 Intron NP_001152851.1
NM_001159380.1 Intron NP_001152852.1
NM_022764.2 Intron NP_073601.2
XM_005256101.1 Intron XP_005256158.1
XM_005256105.1 Intron XP_005256162.1
XM_005256106.1 Intron XP_005256163.1
XM_006721247.3 Intron XP_006721310.1
XM_011523280.2 Intron XP_011521582.1
XM_011523282.2 Intron XP_011521584.1
XM_011523283.2 Intron XP_011521585.1
XM_011523284.2 Intron XP_011521586.1
XM_011523285.1 Intron XP_011521587.1
XM_011523286.1 Intron XP_011521588.1
XM_011523287.1 Intron XP_011521589.1
XM_011523288.1 Intron XP_011521590.1
XM_011523289.1 Intron XP_011521591.1
XM_017023571.1 Intron XP_016879060.1
XM_017023572.1 Intron XP_016879061.1
XM_017023573.1 Intron XP_016879062.1
XM_017023574.1 Intron XP_016879063.1

View Full Product Details