Product Details

SNP ID
rs28636490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20769830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTGGTCTGGTAAAAAGGAGCCA[C/G]CTGGAGGGTGATTGGCTCCTCAGGT
Phenotype
MIM: 611582
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EDDM3B PubMed Links

Gene Details

Gene
EDDM3B
Gene Name
epididymal protein 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022360.4 Intron NP_071755.1

View Full Product Details