Product Details

SNP ID
rs11465569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:102362681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCATGAATTGTAGAGAATTACC[C/G]TTGACCCTTTGGGTGCTTATATCTG
Phenotype
MIM: 604494
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IL18R1 PubMed Links

Gene Details

Gene
IL18R1
Gene Name
interleukin 18 receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282399.1 633 UTR 5 NP_001269328.1
NM_003855.3 633 Silent Mutation CCC,CCG P7P NP_003846.1
XM_005264039.3 633 Silent Mutation CCC,CCG P7P XP_005264096.1
XM_005264040.3 633 Silent Mutation CCC,CCG P7P XP_005264097.1
XM_011512099.1 633 Silent Mutation CCC,CCG P7P XP_011510401.1
XM_017005181.1 633 UTR 5 XP_016860670.1
XM_017005182.1 633 UTR 5 XP_016860671.1
XM_017005183.1 633 UTR 5 XP_016860672.1
XM_017005184.1 633 UTR 5 XP_016860673.1

View Full Product Details