Product Details

SNP ID
rs6509831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53912147 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGAGCATCCAGGGTTCAAGCCCA[A/G]TGCCCTGTGCTCTGGGATCTAGGTC
Phenotype
MIM: 606899 MIM: 176980
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CACNG7 PubMed Links

Gene Details

Gene
CACNG7
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031896.4 Intron NP_114102.2
XM_005259124.3 Intron XP_005259181.1
XM_017027093.1 Intron XP_016882582.1
Gene
PRKCG
Gene Name
protein kinase C gamma
There are no transcripts associated with this gene.

View Full Product Details