Product Details

SNP ID
rs141766791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31702922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTTTGGACAGCGAGCTCAGGTG[G/A]AAGCCAGACTGTGGTGGGCTAAGGA
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CLUHP3 PubMed Links

Gene Details

Gene
CLUHP3
Gene Name
clustered mitochondria homolog pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details