Product Details

SNP ID
rs140480320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47850113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGCAGCACAGAAGGGGCTGAGC[A/G]GGAGCTGTGAAGCCAGGGACCCTAA
Phenotype
MIM: 614966 MIM: 608613
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCRN2 PubMed Links

Gene Details

Gene
SCRN2
Gene Name
secernin 2
There are no transcripts associated with this gene.

Gene
SP6
Gene Name
Sp6 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258248.1 Intron NP_001245177.1
NM_199262.2 Intron NP_954871.1
XM_006722115.3 Intron XP_006722178.1

View Full Product Details