Product Details

SNP ID
rs528539689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:134124007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTAAAGTCTCTAATCTAAGCCAT[A/G]TCCCTATACTGTTTTACATCCTATG
Phenotype
MIM: 601774
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MGAT5 PubMed Links

Gene Details

Gene
MGAT5
Gene Name
mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002410.4 Intron NP_002401.1
XM_005263668.4 Intron XP_005263725.1
XM_005263669.4 Intron XP_005263726.1
XM_005263670.3 Intron XP_005263727.1
XM_006712534.3 Intron XP_006712597.1
XM_011511199.2 Intron XP_011509501.1
XM_011511200.1 Intron XP_011509502.1
XM_011511201.2 Intron XP_011509503.1
XM_011511202.1 Intron XP_011509504.1
XM_011511204.2 Intron XP_011509506.1
XM_017004147.1 Intron XP_016859636.1
XM_017004148.1 Intron XP_016859637.1
XM_017004149.1 Intron XP_016859638.1
Gene
MIR3679
Gene Name
microRNA 3679
There are no transcripts associated with this gene.

View Full Product Details