Product Details

SNP ID
rs10157172
Assay Type
Functionally tested
NCBI dbSNP Submissions
44
Location
Chr.1:20187525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCGCCCATCCCTGTGTGATGG[A/G]TGCTGGGGAGAGCTGGTGTTCCCTC
Phenotype
MIM: 616783
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBXN10 PubMed Links

Gene Details

Gene
UBXN10
Gene Name
UBX domain protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152376.4 Intron NP_689589.1
XM_005245742.3 Intron XP_005245799.1
XM_011540699.2 Intron XP_011539001.1
Gene
UBXN10-AS1
Gene Name
UBXN10 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details