Product Details

SNP ID
rs767164669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:107685803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCATGCGCTCTGCCCTGGCAGCG[A/G]CCCTGTGCAGATCCCTGAGCGTGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PWP1 PubMed Links

Gene Details

Gene
PWP1
Gene Name
PWP1 homolog, endonuclein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317962.1 72 Intron NP_001304891.1
NM_001317963.1 72 UTR 5 NP_001304892.1
NM_007062.2 72 UTR 5 NP_008993.1

View Full Product Details